TCIAT1566

T1566-5G
TCT1566-5GEA 162.31 USD
TCT1566-5G
TCIAT1566
CAS Number: 835-58-5
MDL Number: MFCD00671572
Molecular Formula: C9H5F3O4
Molecular Weight: 234.13
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 151
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR