Perfluoroheptanoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tridecafluoroenanthic acid Tridecafluoroheptanoic acid Tridecafluoroheptylic acid Perfluoroenanthic acid

T1545-5G T1545-25G
TCT1545-25GEA 240.56 USD
TCT1545-25G TCT1545-5G
Perfluoroheptanoic acid ≥98.0% (by titrimetric analysis)
Perfluoroheptanoic acid

CAS Number: 375-85-9
MDL Number: MFCD00039604
Molecular Formula: C7HF13O2
Molecular Weight: 364.06
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 177
Flash Point (°C): 110
Freezing point (°C): 32


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Fluorosurfactants
Other Product Names / Synonyms: Perfluoroenanthic Acid, Perfluoroheptanoic Acid, Tridecafluoroenanthic Acid

Formula: C₇HF₁₃O₂
Boiling Pt: 177 °C
Flash Pt: 110 °C
MDL Number: MFCD00039604
CAS Number: 375-85-9
UN: 3261
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/55/57/28995557.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR