Trimethylsilylacetylene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethynyltrimethylsilane

T1239-5ML T1239-25ML T1239-250ML
TCT1239-25MLEA 156.28 USD
TCT1239-25ML TCT1239-5ML TCT1239-250ML
Trimethylsilylacetylene ≥98.0% (by GC)
Trimethylsilylacetylene

CAS Number: 1066-54-2
MDL Number: MFCD00008569
Molecular Formula: C5H10Si
Molecular Weight: 98.22
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 53
Flash Point (°C): -34
Specific Gravity (20/20): 0.71


Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis), Terminal Alkynes (Click Chemistry), Click Chemistry, Materials Chemistry, Langmuir-Blodgett(LB) Film Forming Agents
Other Product Names / Synonyms: Ethynyltrimethylsilane

Formula: C₅H₁₀Si
MW: 98.22 g/mol
Boiling Pt: 53 °C
Density: 0.71 (20 °C)
Flash Pt: –34 °C
MDL Number: MFCD00008569
CAS Number: 1066-54-2
https://us.vwr.com/stibo/web/std.lang.all/52/79/28995279.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR