Thiophen-3-acetonitrile ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 3-Thiopheneacetonitrile2-(Thiophen-3-yl)acetonitrile

T1193-5G
TCT1193-5GEA 79 USD
TCT1193-5G
Thiophen-3-acetonitrile ≥97.0% (by GC)
Thiophen-3-acetonitrile

CAS Number: 13781-53-8
MDL Number: MFCD00005471
Molecular Formula: C6H5NS
Molecular Weight: 123.17
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 125
Flash Point (°C): 119
Specific Gravity (20/20): 1.16

Formula: C₆H₅NS
MW: 123.18 g/mol
Boiling Pt: 125 °C
Density: 1.16 (20 °C)
Flash Pt: 119 °C
MDL Number: MFCD00005471
UN: 3276
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/52/41/28995241.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR