Borane trimethylamine complex ≥97.0% (by titrimetric analysis)
Supplier: TCI America
T1181-25G
TCT1181-25GEA
100.45
USD
TCT1181-25G
Borane trimethylamine complex ≥97.0% (by titrimetric analysis)
Borane trimethylamine complex
CAS Number: 75-22-9
MDL Number: MFCD00012420
Molecular Formula: C3H12BN
Molecular Weight: 72.95
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 95
Flash Point (°C): 64
Product Categories: Synthetic Organic Chemistry, Reduction, Boration
Other Product Names / Synonyms: Borane - Trimethylamine Complex, TMAB
Formula:
C₃H₁₂BN Melting Pt: 95 °C Flash Pt: 64 °C |
MDL Number:
MFCD00012420 CAS Number: 75-22-9 UN: 1325 ADR: 4.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...