Chloromethyl thiocyanate

T1179-5G T1179-25G
TCT1179-25GEA 79.56 USD
TCT1179-25G TCT1179-5G
Chloromethyl thiocyanate
Chloromethyl thiocyanate

CAS Number: 3268-79-9
MDL Number: MFCD00001831
Molecular Formula: C2H2ClNS
Molecular Weight: 107.56
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 95
Flash Point (°C): 83
Specific Gravity (20/20): 1.37


Other Product Names / Synonyms: Thiocyanic Acid Chloromethyl Ester

Formula: C₂H₂ClNS
Boiling Pt: 95 °C
Density: 1.37 (20 °C)
Flash Pt: 83 °C
MDL Number: MFCD00001831
CAS Number: 3268-79-9
UN: 2810
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/52/29/28995229.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR