Tetramethylgermane ≥98.0% (by GC)
Supplier: TCI America
T1158-5G
T1158-1G
TCT1158-1GEA
65.98
USD
TCT1158-1G
TCT1158-5G
Tetramethylgermane ≥98.0% (by GC)
Tetramethylgermane
CAS Number: 865-52-1
MDL Number: MFCD00014843
Molecular Formula: C4H12Ge
Molecular Weight: 132.77
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 43
Flash Point (°C): -37
Specific Gravity (20/20): 0.97
Formula:
C₄H₁₂Ge MW: 132.75 g/mol Boiling Pt: 43 °C Density: 0.97 (20 °C) Flash Pt: –37 °C |
MDL Number:
MFCD00014843 CAS Number: 865-52-1 UN: 1993 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...