2,3,4-Trimethyl-2-pentene ≥98.0% (by GC)

Supplier: TCI America

T1080-1ML T1080-0.1ML
TCT1080-1MLEA 86.24 USD
TCT1080-1ML TCT1080-0.1ML
2,3,4-Trimethyl-2-pentene ≥98.0% (by GC)
2,3,4-Trimethyl-2-pentene

CAS Number: 565-77-5
MDL Number: MFCD00059214
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 116
Flash Point (°C): 12
Specific Gravity (20/20): 0.74

https://us.vwr.com/stibo/web/std.lang.all/51/52/28995152.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR