2,4,4-Trimethyl-2-oxazoline ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,4,4-Trimethyl-4,5-dihydrooxazole

T1060-25ML
TCT1060-25MLEA 96 USD
TCT1060-25ML
2,4,4-Trimethyl-2-oxazoline ≥98.0% (by GC)
2,4,4-Trimethyl-2-oxazoline

CAS Number: 1772-43-6
MDL Number: MFCD00005301
Molecular Formula: C6H11NO
Molecular Weight: 113.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 112
Flash Point (°C): 13
Specific Gravity (20/20): 0.92

Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 112 °C
Density: 0.92 (20 °C)
Flash Pt: 13 °C
MDL Number: MFCD00005301
CAS Number: 1772-43-6
https://us.vwr.com/stibo/web/std.lang.all/51/34/28995134.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR