2,4,6-Trifluoroaniline ≥98.0% (by GC)

Supplier: TCI America

T1038-5G
TCT1038-5GEA 190 USD
TCT1038-5G
2,4,6-Trifluoroaniline ≥98.0% (by GC)
2,4,6-Trifluoroaniline

CAS Number: 363-81-5
MDL Number: MFCD00007650
Molecular Formula: C6H4F3N
Molecular Weight: 147.10
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Flash Point (°C): 57
Freezing point (°C): 33

Formula: C₆H₄F₃N
MW: 147.1 g/mol
Flash Pt: 57 °C
MDL Number: MFCD00007650
CAS Number: 363-81-5
UN: 2926
ADR: 4.1,III
https://us.vwr.com/stibo/web/std.lang.all/51/13/28995113.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR