Tetraethylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
Tetrylammonium tetrafluoroborate
T0983-25G
TCT0983-25GEA
73.05
USD
TCT0983-25G
Tetraethylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Tetraethylammonium tetrafluoroborate
CAS Number: 429-06-1
MDL Number: MFCD00011827
Molecular Formula: C8H20BF4N
Molecular Weight: 217.06
Purity/Analysis Method:
Form: Crystal
Color: White
Formula:
C₈H₂₀BF₄N MW: 217.06 g/mol |
MDL Number:
MFCD00011827 CAS Number: 429-06-1 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...