3-(Trifluoromethyl)phenyltrimethylammonium hydroxide 5% in methanol
Supplier: TCI America
Synonyms:
m-TFPTAH
[for Transesterification of Glyceride]
CAS Number: 68254-41-1
MDL Number: MFCD00059473
Molecular Formula: C10H14F3NO
Molecular Weight: 221.22
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 11
Storage Temperature: 0-10°C
Other Product Names / Synonyms: m-TFPTAH, Trimethyl-3-trifluoromethylphenylammonium Hydroxide, Trimethyl-alpha,alpha,alpha-trifluoro-m-tolylammonium Hydroxide
Formula:
C₁₀H₁₄F₃NO MW: 221.22 g/mol Flash Pt: 11 °C |
MDL Number:
MFCD00059473 CAS Number: 68254-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...