Azidotrimethylsilane ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
Trimethylsilyl azide
T0801-25G
TCT0801-25GEA
127.12
USD
TCT0801-25G
Azidotrimethylsilane ≥95.0% (by GC)
Azidotrimethylsilane
CAS Number: 4648-54-8
MDL Number: MFCD00001986
Molecular Formula: C3H9N3Si
Molecular Weight: 115.21
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 97
Flash Point (°C): 30
Specific Gravity (20/20): 0.88
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis), Click Chemistry
Other Product Names / Synonyms: Azidotrimethylsilane
Formula:
C₃H₉N₃Si Boiling Pt: 97 °C Density: 0.88 (20 °C) Flash Pt: 30 °C |
MDL Number:
MFCD00001986 CAS Number: 4648-54-8 UN: 1992 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...