(±)-Tetrahydrofurfuryl chloride ≥98.0% (by GC)

Supplier: TCI America

T0800-25G
TCT0800-25GEA 62.9 USD
TCT0800-25G
(±)-Tetrahydrofurfuryl chloride ≥98.0% (by GC)
(±)-Tetrahydrofurfuryl chloride

CAS Number: 3003-84-7
MDL Number: MFCD00005370
Molecular Formula: C5H9ClO
Molecular Weight: 120.58
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 151
Flash Point (°C): 45
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: 2-(Chloromethyl)tetrahydrofuran

Formula: C₅H₉ClO
MW: 120.58
Boiling Pt: 151 °C
Density: 1.11 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00005370
CAS Number: 3003-84-7
https://us.vwr.com/stibo/web/std.lang.all/49/21/28994921.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR