1-(2-Methylphenyl)-2-thiourea ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 1-(2-Tolyl)thiourea 2-Methylphenylthiourea N-(2-Methylphenyl)thiourea o-Tolylthiourea

T0657-25G
TCT0657-25GEA 82.22 USD
TCT0657-25G
1-(2-Methylphenyl)-2-thiourea ≥98.0% (by HPLC)
1-(2-Methylphenyl)-2-thiourea

CAS Number: 614-78-8
MDL Number: MFCD00041162
Molecular Formula: C8H10N2S
Molecular Weight: 166.24
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 156

Formula: (C₆H₄(CH₃))NC(S)NH₂
MW: 166.25 g/mol
Melting Pt: 156 °C
MDL Number: MFCD00041162
CAS Number: 614-78-8
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/48/31/28994831.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR