1,3,5-Trinitrobenzene ≥98.0% (by anhydrous basis) wetted with ca. 40% water

Supplier: TCI America

T0496-10G
TCT0496-10GEA 216.6 USD
TCT0496-10G
1,3,5-Trinitrobenzene ≥98.0% (by anhydrous basis) wetted with ca. 40% water
1,3,5-Trinitrobenzene

CAS Number: 99-35-4
MDL Number: MFCD00059179
Molecular Formula: C6H3N3O6
Molecular Weight: 213.11
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 122

Formula: C₆H₃N₃O₆
MW: 213.11 g/mol
Melting Pt: 122 °C
CAS Number: 99-35-4
ADR: 1,

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR