Bromal (tribromoacetaldehyde) ≥96.0% (by GC)

Supplier: TCI America

Synonyms: BromalTribromoacetaldehyde

T0350-25G
TCT0350-25GEA 43.64 USD
TCT0350-25G
Bromal (tribromoacetaldehyde) ≥96.0% (by GC)
Bromal (tribromoacetaldehyde)

CAS Number: 115-17-3
MDL Number: MFCD00006961
Molecular Formula: C2HBr3O
Molecular Weight: 280.74
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 174
Flash Point (°C): 65
Specific Gravity (20/20): 2.68


Other Product Names / Synonyms: Tribromoacetaldehyde

https://us.vwr.com/stibo/web/std.lang.all/46/34/28994634.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR