2-Thienylacetic acid ≥96.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Thiophene-2-acetic acid2-(Thiophen-2-yl)acetic acid2-Thiopheneacetic acid

T0217-25G
TCT0217-25GEA 352 USD
TCT0217-25G
2-Thienylacetic acid ≥96.0% (by GC, titration analysis)
2-Thienylacetic acid

CAS Number: 1918-77-0
MDL Number: MFCD00005458
Molecular Formula: C6H6O2S
Molecular Weight: 142.17
Purity/Analysis Method: 96.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 160
Melting point (°C): 63


Other Product Names / Synonyms: 2-Thienylacetic Acid

Formula: C₆H₆O₂S
MW: 142.18 g/mol
Boiling Pt: 160 °C
Melting Pt: 63 °C
MDL Number: MFCD00005458
CAS Number: 1918-77-0
UN: 3261
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/45/28/28994528.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR