Thiobenzamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Benzothioamide

T0194-25G T0194-100G
TCT0194-100GEA 274.05 USD
TCT0194-100G TCT0194-25G
Thiobenzamide ≥98.0% (by HPLC, total nitrogen)
Thiobenzamide

CAS Number: 2227-79-4
MDL Number: MFCD00008060
Molecular Formula: C7H7NS
Molecular Weight: 137.20
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 117

Formula: C₇H₇NS
MW: 137.21 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00008060
CAS Number: 2227-79-4
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/45/11/28994511.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR