1,3,6,8-Tetranitrocarbazole ≥70.0% (by HPLC on dried basis) wetted with ca. 40% water

T0159-25G
TCT0159-25GEA 108.15 USD
TCT0159-25G
1,3,6,8-Tetranitrocarbazole ≥70.0% (by HPLC on dried basis) wetted with ca. 40% water
1,3,6,8-Tetranitrocarbazole

CAS Number: 4543-33-3
MDL Number: MFCD00022211
Molecular Formula: C12H5N5O8
Molecular Weight: 347.20
Purity/Analysis Method: 70.0% (HPLC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 291

Formula: C₁₂H₅N₅O₈
Melting Pt: 291 °C
MDL Number: MFCD00022211
CAS Number: 4543-33-3
https://us.vwr.com/stibo/web/std.lang.all/44/82/28994482.jpg

Order Now

Specification Test Results

Purity/Analysis Method 70.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR