1-Tetradecanol ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1-Tetradecyl alcohol
CAS Number: 112-72-1
MDL Number: MFCD00004757
Molecular Formula: C14H30O
Molecular Weight: 214.39
Purity/Analysis Method:
Form: Crystal
Color: White
Boiling point (°C): 173
Melting point (°C): 38
Flash Point (°C): 156
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Myristyl Alcohol
Formula:
H₃C(CH₂)₁₂CH₂OH MW: 214.39 g/mol Boiling Pt: 173 °C Melting Pt: 38 °C Flash Pt: 156 °C |
MDL Number:
MFCD00004757 CAS Number: 112-72-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...