DL-Sodium lactate 60% in water
Supplier: TCI America
Synonyms:
(±)-2-Hydroxypropanoic acid sodium salt
S0928-500G
TCS0928-500GEA
65.52
USD
TCS0928-500G
DL-Sodium lactate 60% in water
DL-Sodium lactate
CAS Number: 72-17-3
MDL Number: MFCD00065400
Molecular Formula: C3H6O3
Molecular Weight: 112.06
Form: Clear Liquid
Color: Colorless
Other Product Names / Synonyms: DL-Lactic Acid Sodium Salt, Sodium 2-Hydroxypropionate, 2-Hydroxypropionic Acid Sodium Salt
Formula:
H₃CCH(OH)COONa MW: 112.06 g/mol |
MDL Number:
MFCD00065400 CAS Number: 72-17-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...