N-(2-Hydroxy-3-sulfopropyl)-3,5-dimethoxyaniline sodium salt ≥98.0% (by HPLC) for biochemical research
Supplier: TCI America
Synonyms:
3-(3,5-Dimethoxyanilino)-2-hydroxypropanesulfonic Acid Sodium Salt
[for Biochemical Research]
CAS Number: 82692-88-4
MDL Number: MFCD00063425
Molecular Formula: C11H17NO6S
Molecular Weight: 313.30
Purity/Analysis Method:
Form: Crystal
Color: White
Storage Temperature: 0-10°C
Product Categories: Bioscience, Peroxidase Substrates, Trinder's Reagents, ROS/RNS Detection Reagents, Redox Research Reagents
Other Product Names / Synonyms: 3-(3,5-Dimethoxyanilino)-2-hydroxypropanesulfonic Acid Sodium Salt, Sodium 3-(3,5-Dimethoxyanilino)-2-hydroxypropanesulfonate, HDAOS
Formula:
C₁₁H₁₇NO₆S MW: 313.30 g/mol |
MDL Number:
MFCD00063425 CAS Number: 82692-88-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...