Sodium ethoxide ∼20% in ethanol

Supplier: TCI America

Synonyms: Sodium ethylateSodium ethanolate

S0548-500ML
TCS0548-500MLEA 65.49 USD InStock
TCS0548-500ML
Sodium ethoxide ∼20% in ethanol
Sodium ethoxide

CAS Number: 141-52-6
MDL Number: MFCD00012417
Molecular Formula: C2H5NaO
Molecular Weight: 68.05
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 9
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Sodium Ethylate

Formula: C₂H₅NaO
MW: 68,05 g/mol
Melting Pt: 260 °C
Density: 0,88 (20 °C)
Flash Pt: 9 °C
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 3206
ADR: 4.2,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR