Sodium ethoxide ≥95.0% (by titrimetric analysis)
Supplier: TCI America
|
Synonyms:
Sodium ethylate
, Sodium ethanolate
S0547-100G
TCS0547-100GEA
42.78
USD
TCS0547-100G
Sodium ethoxide ≥95.0% (by titrimetric analysis)
Sodium ethoxide
CAS Number: 141-52-6
MDL Number: MFCD00012417
Molecular Formula: C2H5NaO
Molecular Weight: 68.05
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 260
Flash Point (°C): 30
Other Product Names / Synonyms: Sodium Ethylate
Formula:
C₂H₅NaO MW: 68.05 g/mol Melting Pt: 260 °C Flash Pt: 30 °C |
MDL Number:
MFCD00012417 CAS Number: 141-52-6 UN: 3206 ADR: 4.2,II |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...