(R)-(+)-Epoxystyrene ≥96.0% (by GC)

Supplier: TCI America

Synonyms: (R)-Styrene oxide(R)-(+)-Styrene oxide

S0516-5G S0516-25G
TCS0516-25GEA 247.33 USD
TCS0516-25G TCS0516-5G
(R)-(+)-Epoxystyrene ≥96.0% (by GC)
(R)-(+)-Epoxystyrene

CAS Number: 20780-53-4
MDL Number: MFCD00066210
Molecular Formula: C8H8O
Molecular Weight: 120.15
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 194
Flash Point (°C): 77
Specific Gravity (20/20): 1.05
Specific rotation [a]20/D: 33 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-1,2-Epoxyethylbenzene, (R)-Phenylethylene Oxide, (R)-Phenyloxirane

Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 194 °C
Density: 1.05 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00066210
CAS Number: 20780-53-4
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/41/62/28994162.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR