TCIAS0461

S0461-10G S0461-1G
76220-472EA 630.79 USD
76220-472 TCS0461-1G
TCIAS0461
CAS Number: 90-39-1
MDL Number: MFCD00069653
Molecular Formula: C15H26N2
Molecular Weight: 234.39
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 147
Flash Point (°C): 113
Specific Gravity (20/20): 1.02
Specific rotation [a]20/D: -18 deg (C=1, EtOH)
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: Lupinidine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR