1-Heptene ≥99.5% (by GC)
Supplier: TCI America
S0337-5ML
TCS0337-5MLEA
112.67
USD
TCS0337-5ML
1-Heptene ≥99.5% (by GC)
1-Heptene
[Standard Material for GC]
CAS Number: 592-76-7
MDL Number: MFCD00009531
Molecular Formula: C7H14
Molecular Weight: 98.19
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 94
Flash Point (°C): -4
Specific Gravity (20/20): 0.70
Formula:
C₇H₁₄ MW: 98.19 g/mol Boiling Pt: 94 °C Density: 0.70 (20 °C) Flash Pt: –4 °C |
MDL Number:
MFCD00009531 CAS Number: 592-76-7 UN: 2278 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...