Methyl propionate ≥99.5% (by GC)
Supplier: TCI America
Synonyms:
Propionic acid methyl ester
[Standard Material for GC]
CAS Number: 554-12-1
MDL Number: MFCD00009306
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 80
Melting point (°C): -88
Flash Point (°C): -4
Specific Gravity (20/20): 0.92
Product Categories: Analytical Science, Fatty Acid Methyl Esters (GC Standard)
Other Product Names / Synonyms: Propionic Acid Methyl Ester
Formula:
C₄H₈O₂ MW: 88.11 g/mol Boiling Pt: 80 °C Melting Pt: –88 °C Density: 0.92 (20 °C) Flash Pt: –4 °C |
MDL Number:
MFCD00009306 CAS Number: 554-12-1 UN: 1248 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...