[RuCl(p-cymene)((R)-dtbm-segphos®)]Cl
Supplier: TCI America
Synonyms:
Chloro[(R)-(-)-5,5'-bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
R0158-1G
R0158-200MG
TCR0158-1GEA
199
USD
TCR0158-1G
TCR0158-200MG
[RuCl(p-cymene)((R)-dtbm-segphos®)]Cl
[RuCl(p-cymene)((R)-dtbm-segphos®)]Cl
CAS Number: 944451-32-5
MDL Number: MFCD09753019
Molecular Formula: C84H114Cl2O8P2Ru
Molecular Weight: 1485.75
Form: Crystal
Color: Yellow
Other Product Names / Synonyms: Chloro[(R)-(-)-5,5'-bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
Formula:
C₈₄H₁₁₄Cl₂O₈P₂Ru MW: 1485.75 g/mol |
MDL Number:
MFCD09753019 CAS Number: 944451-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...