Rabeprazole sodium salt ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Sodium 2-(((4-(3-methoxypropoxy)-3-methylpyridin-2-yl)methyl)sulfinyl)benzo[d]imidazol-1-ideSodium 2-(((4-(3-methoxypropoxy)-3-methylpyridin-2-yl)methyl)sulphinyl)benzo[d]imidazol-1-ide

R0115-1G R0115-100MG
TCR0115-100MGEA 140.68 USD
TCR0115-100MG TCR0115-1G
Rabeprazole sodium salt ≥98.0% (by HPLC)
Rabeprazole sodium salt

CAS Number: 117976-90-6
MDL Number: MFCD02092688
Molecular Formula: C18H21N3O3S
Molecular Weight: 381.43
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 150
Storage Temperature: 0°C

Formula: C₁₈H₂₁N₃O₃S
MW: 381.43 g/mol
Melting Pt: 150 °C
MDL Number: MFCD02092688
CAS Number: 117976-90-6
https://us.vwr.com/stibo/web/std.lang.all/38/10/28993810.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR