Raloxifene hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
[6-Hydroxy-2-(4-hydroxyphenyl)benzo[b]thiophen-3-yl][4-[2-(1-piperidinyl)ethoxy]phenyl]methanone Hydrochloride
CAS Number: 82640-04-8
MDL Number: MFCD01938233
Molecular Formula: C28H27NO4S
Molecular Weight: 510.05
Purity/Analysis Method:
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 263
Lambda max.: 286 nm
Product Categories: Bioscience, Antitumor Ingredients, Anti-osteoporosis Ingredients
Other Product Names / Synonyms: [6-Hydroxy-2-(4-hydroxyphenyl)benzo[b]thiophen-3-yl][4-[2-(1-piperidinyl)ethoxy]phenyl]methanone Hydrochloride
Formula:
C₂₈H₂₇NO₄S MW: 510.05 g/mol Melting Pt: 263 °C |
MDL Number:
MFCD01938233 CAS Number: 82640-04-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...