5-Quinolinecarboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Q0089-1G
TCQ0089-1GEA 131 USD
TCQ0089-1G
5-Quinolinecarboxylic acid ≥98.0% (by HPLC, titration analysis)
5-Quinolinecarboxylic acid

CAS Number: 7250-53-5
MDL Number: MFCD00047608
Molecular Formula: C10H7NO2
Molecular Weight: 173.17
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 342

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Melting Pt: 342 °C
MDL Number: MFCD00047608
CAS Number: 7250-53-5

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR