3-(1,3-Dioxoisoindolin-2-yl)propanoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Phthalimidopropionic acid3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

P2101-5G P2101-25G
TCP2101-25GEA 207 USD
TCP2101-25G TCP2101-5G
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid ≥98.0% (by HPLC, titration analysis)
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid

CAS Number: 3339-73-9
MDL Number: MFCD00023096
Molecular Formula: C11H9NO4
Molecular Weight: 219.20
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 153


Other Product Names / Synonyms: N-Phthaloyl-beta-alanine

Formula: C₁₁H₉NO₄
MW: 219.196 g/mol
Melting Pt: 153 °C
MDL Number: MFCD00023096
CAS Number: 3339-73-9
https://us.vwr.com/stibo/web/std.lang.all/33/13/28993313.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR