6-Phenyl-3(2H)-pyridazinone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 6-Phenylpyridazin-3(2H)-one3-Hydroxy-6-phenylpyridazine

P2060-1G P2060-5G
TCP2060-1GEA 142 USD
TCP2060-1G TCP2060-5G
6-Phenyl-3(2H)-pyridazinone ≥98.0% (by GC)
6-Phenyl-3(2H)-pyridazinone

CAS Number: 2166-31-6
MDL Number: MFCD00114953
Molecular Formula: C10H8N2O
Molecular Weight: 172.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 203
Lambda max.: 249 nm (HCl aq.)


Other Product Names / Synonyms: 3-Hydroxy-6-phenylpyridazine

Formula: C₁₀H₈N₂O
MW: 172.19 g/mol
Melting Pt: 203 °C
MDL Number: MFCD00114953
CAS Number: 2166-31-6
https://us.vwr.com/stibo/web/std.lang.all/32/85/28993285.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR