(Chloromethyl)pentamethylbenzene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2,3,4,5,6-Pentamethylbenzyl chloride
, Pentamethylbenzyl chloride
P2046-5G
P2046-25G
TCP2046-5GEA
315
USD
TCP2046-5G
TCP2046-25G
(Chloromethyl)pentamethylbenzene ≥98.0% (by GC)
(Chloromethyl)pentamethylbenzene
CAS Number: 484-65-1
MDL Number: MFCD00000900
Molecular Formula: C12H17Cl
Molecular Weight: 196.72
Purity/Analysis Method:
Form: Crystal
Color: White
Boiling point (°C): 163
Melting point (°C): 82
Formula:
(C₆(CH₃)₅)CH₂Cl MW: 196.72 g/mol Boiling Pt: 163 °C Melting Pt: 82 °C |
MDL Number:
MFCD00000900 CAS Number: 484-65-1 UN: 3261 ADR: 8,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...