Phenylmagnesium bromide 16% ∼1 mol/L in THF
Supplier: TCI America
P2025-250G
TCP2025-250GEA
180.08
USD
TCP2025-250G
Phenylmagnesium bromide 16% ∼1 mol/L in THF
Phenylmagnesium bromide
CAS Number: 100-58-3
MDL Number: MFCD00000038
Molecular Formula: C6H5BrMg
Molecular Weight: 181.32
Flash Point (°C): -17
Formula:
C₆H₅BrMg Flash Pt: –17 °C |
MDL Number:
MFCD00000038 CAS Number: 100-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...