[6,6]-Phenyl-C61-butyric acid dodecyl ester ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: [60]PCB-C12

P2015-100MG
TCP2015-100MGEA 424 USD
TCP2015-100MG
[6,6]-Phenyl-C61-butyric acid dodecyl ester ≥98.0% (by HPLC)
[6,6]-Phenyl-C61-butyric acid dodecyl ester

CAS Number: 571177-69-0
Molecular Formula: C83H36O2
Molecular Weight: 1065.20
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Color: Black


Product Categories: Materials Chemistry, Solar Cell Research Reagents, Fullerenes (Nanocarbon Materials)
Other Product Names / Synonyms: [60]PCB-C12

Formula: C₈₃H₃₆O₂
MW: 1065.20 g/mol
CAS Number: 571177-69-0
https://us.vwr.com/stibo/web/std.lang.all/32/54/28993254.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR