[6,6]-Phenyl-C61-butyric acid n-octyl ester ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: PCBO[60]PCB-C8

P2014-100MG
TCP2014-100MGEA 410 USD
TCP2014-100MG
[6,6]-Phenyl-C61-butyric acid n-octyl ester ≥98.0% (by HPLC)
[6,6]-Phenyl-C61-butyric acid n-octyl ester

CAS Number: 571177-68-9
MDL Number: MFCD09265175
Molecular Formula: C79H28O2
Molecular Weight: 1009.09
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Color: Black


Product Categories: Materials Chemistry, Solar Cell Research Reagents, Fullerenes (Nanocarbon Materials)
Other Product Names / Synonyms: PCBO, [60]PCB-C8

Formula: C₇₉H₂₈O₂
MW: 1009.09 g/mol
MDL Number: MFCD09265175
CAS Number: 571177-68-9
https://us.vwr.com/stibo/web/std.lang.all/32/53/28993253.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR