[6,6]-Phenyl-C61-butyric acid butyl ester ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: PCBB[60]PCB-C4

P2013-100MG
TCP2013-100MGEA 264 USD
TCP2013-100MG
[6,6]-Phenyl-C61-butyric acid butyl ester ≥98.0% (by HPLC)
[6,6]-Phenyl-C61-butyric acid butyl ester

CAS Number: 571177-66-7
MDL Number: MFCD09842755
Molecular Formula: C75H20O2
Molecular Weight: 952.98
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Color: Black


Product Categories: Materials Chemistry, Solar Cell Research Reagents, Fullerenes (Nanocarbon Materials)
Other Product Names / Synonyms: PCBB, [60]PCB-C4

Formula: C₇₅H₂₀O₂
MW: 952.98 g/mol
MDL Number: MFCD09842755
CAS Number: 571177-66-7
https://us.vwr.com/stibo/web/std.lang.all/32/52/28993252.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR