(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Bis(tricyclohexylphosphine)-3-phenyl-1H-inden-1-ylideneruthenium(II) dichloride
P2004-1G
TCP2004-1GEA
211
USD
TCP2004-1G
(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran ≥97.0% (by titrimetric analysis)
(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran
CAS Number: 250220-36-1
MDL Number: MFCD07369034
Molecular Formula: C51H76Cl2P2Ru
Molecular Weight: 923.09
Purity/Analysis Method:
Form: Crystal
Color: Deep Yellow
Formula:
C₅₁H₇₆Cl₂P₂Ru MW: 923.09 g/mol |
MDL Number:
MFCD07369034 CAS Number: 250220-36-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...