3-(3-Pyridyl)propionic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Pyridinepropionic acid3-Pyridinepropionic Acid

P1992-1G P1992-5G
TCP1992-1GEA 53.8 USD
TCP1992-1G TCP1992-5G
3-(3-Pyridyl)propionic acid ≥98.0% (by GC, titration analysis)
3-(3-Pyridyl)propionic acid

CAS Number: 3724-19-4
MDL Number: MFCD00274197
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 158


Other Product Names / Synonyms: 3-Pyridinepropionic Acid

Formula: C₈H₉NO₂
MW: 151.17 g/mol
Melting Pt: 158 °C
MDL Number: MFCD00274197
CAS Number: 3724-19-4
https://us.vwr.com/stibo/web/std.lang.all/32/35/28993235.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR