1-Phenylcyclohexanol ≥98.0% (by GC)
Supplier: TCI America
P1988-25G
P1988-5G
TCP1988-5GEA
94.4
USD
TCP1988-5G
TCP1988-25G
1-Phenylcyclohexanol ≥98.0% (by GC)
1-Phenylcyclohexanol
CAS Number: 1589-60-2
MDL Number: MFCD00021393
Molecular Formula: C12H16O
Molecular Weight: 176.26
Purity/Analysis Method:
Form: Crystal
Color: White
Boiling point (°C): 100
Melting point (°C): 63
Other Product Names / Synonyms: 1-Hydroxy-1-phenylcyclohexane, 1-Phenyl-1-cyclohexanol
Formula:
C₁₂H₁₆O MW: 176.26 g/mol Boiling Pt: 100 °C Melting Pt: 63 °C |
MDL Number:
MFCD00021393 CAS Number: 1589-60-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...