(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: D-(+)-3-Phenyllactic acidD(+)-Phenyllactic acid(R)-2-Hydroxy-3-phenylpropanoic acid

P1981-5G P1981-1G
TCP1981-1GEA 41.62 USD
TCP1981-1G TCP1981-5G
(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥98.0% (by HPLC, titration analysis)
(R)-(+)-2-Hydroxy-3-phenylpropionic acid

CAS Number: 7326-19-4
MDL Number: MFCD00078062
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 125
Specific rotation [a]20/D: 22 deg (C=2, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(+)-2-Hydroxy-3-phenylpropionic Acid, (R)-(+)-3-Phenyllactic Acid

Formula: C₉H₁₀O₃
MW: 166.1739 g/mol
Melting Pt: 125 °C
MDL Number: MFCD00078062
CAS Number: 7326-19-4
https://us.vwr.com/stibo/web/std.lang.all/32/28/28993228.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR