Probenecid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-[(dipropylamino)sulphonyl]benzoic acid

P1975-25G
TCP1975-25GEA 74.64 USD
TCP1975-25G
Probenecid ≥98.0% (by HPLC, titration analysis)
Probenecid

CAS Number: 57-66-9
MDL Number: MFCD00038402
Molecular Formula: C13H19NO4S
Molecular Weight: 285.36
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 200


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 4-(N,N-Dipropylsulfamoyl)benzoic Acid

Formula: C₁₃H₁₉NO₄S
MW: 285.4 g/mol
Melting Pt: 200 °C
MDL Number: MFCD00038402
CAS Number: 57-66-9
https://us.vwr.com/stibo/web/std.lang.all/32/24/28993224.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR