Phenylsulfur pentafluoride ≥98.0% (by GC)

Supplier: TCI America

P1969-1G
TCP1969-1GEA 149.16 USD
TCP1969-1G
Phenylsulfur pentafluoride ≥98.0% (by GC)
Phenylsulphur pentafluoride

CAS Number: 2557-81-5
MDL Number: MFCD09263740
Molecular Formula: C6H5F5S
Molecular Weight: 204.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 149
Melting point (°C): -10
Flash Point (°C): 48
Specific Gravity (20/20): 1.51

Formula: C₆H₅F₅S
Boiling Pt: 149 °C
Melting Pt: –10 °C
Density: 1.51 (20 °C)
Flash Pt: 48 °C
MDL Number: MFCD09263740
CAS Number: 2557-81-5
https://us.vwr.com/stibo/web/std.lang.all/32/19/28993219.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR