2-(Phosphonooxy)benzoic acid ≥97.0% (by HPLC, titration analysis) for biochemical research

Synonyms: 2-Carboxyphenyl PhosphateFosfosal

P1952-1G
TCP1952-1GEA 69.93 USD
TCP1952-1G
2-(Phosphonooxy)benzoic acid ≥97.0% (by HPLC, titration analysis) for biochemical research
2-(Phosphonooxy)benzoic acid

[for Biochemical Research]
CAS Number: 6064-83-1
MDL Number: MFCD00042645
Molecular Formula: C7H7O6P
Molecular Weight: 218.10
Purity/Analysis Method: 97.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 161
Storage Temperature: 0-10°C


Product Categories: Bioscience, Alkali Phosphatase Substrates
Other Product Names / Synonyms: 2-Carboxyphenyl Phosphate, Fosfosal

Formula: C₇H₇O₆P
MW: 218.10 g/mol
Melting Pt: 161 °C
MDL Number: MFCD00042645
CAS Number: 6064-83-1
https://us.vwr.com/stibo/web/std.lang.all/32/04/28993204.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR