Pyridazine-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Pyridazinecarboxylic acid

P1907-1G
TCP1907-1GEA 270 USD
TCP1907-1G
Pyridazine-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Pyridazine-4-carboxylic acid

CAS Number: 50681-25-9
MDL Number: MFCD00011576
Molecular Formula: C5H4N2O2
Molecular Weight: 124.10
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 238

Formula: C₅H₄N₂O₂
MW: 124.10 g/mol
Melting Pt: 238 °C
MDL Number: MFCD00011576
CAS Number: 50681-25-9
https://us.vwr.com/stibo/web/std.lang.all/31/82/28993182.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR