2-(1H-Pyrazol-5-yl)aniline ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(1H-Pyrazol-3-yl)aniline3-(2-Aminophenyl)-1H-pyrazole

P1902-1G
TCP1902-1GEA 513 USD
TCP1902-1G
2-(1H-Pyrazol-5-yl)aniline ≥97.0% (by GC, titration analysis)
2-(1H-Pyrazol-5-yl)aniline

CAS Number: 111562-32-4
MDL Number: MFCD09028476
Molecular Formula: C9H9N3
Molecular Weight: 159.19
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 129


Product Categories: Synthetic Organic Chemistry, C-H Bond Activation Reaction, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: 5-(2-Aminophenyl)pyrazole

Formula: C₉H₉N₃
MW: 159.1879 g/mol
Melting Pt: 129 °C
MDL Number: MFCD09028476
CAS Number: 111562-32-4
https://us.vwr.com/stibo/web/std.lang.all/31/77/28993177.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR