Pyrrole-3-carboxylic acid ≥95.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
1H-Pyrrole-3-carboxylic acid
P1899-1G
P1899-5G
TCP1899-1GEA
107.59
USD
TCP1899-1G
TCP1899-5G
Pyrrole-3-carboxylic acid ≥95.0% (by GC, titration analysis)
Pyrrole-3-carboxylic acid
CAS Number: 931-03-3
MDL Number: MFCD00800594
Molecular Formula: C5H5NO2
Molecular Weight: 111.10
Purity/Analysis Method:
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 148
Formula:
C₅H₅NO₂ MW: 111.10 g/mol Melting Pt: 148 °C |
MDL Number:
MFCD00800594 CAS Number: 931-03-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...